Anti-SKA3 Goat Polyclonal Antibody

Lieferant: Abnova
PAB17243
ABNOPAB17243EA 523 EUR
ABNOPAB17243
Anti-SKA3 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of C13orf3.

Recommended Dilutions: ELISA: 1:64000

Type: Primary
Antigen: SKA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR