Anti-RAD9 Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB17290
ABNOPAB17290EA 423 EUR
ABNOPAB17290
Anti-RAD9 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of RAD9A.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: RAD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR