Anti-H4 Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB17307
ABNOPAB17307EA 423 EUR
ABNOPAB17307
Anti-H4 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of Histone H4.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: H4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR