Anti-GK3P Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB17384
ABNOPAB17384EA 423 EUR
ABNOPAB17384
Anti-GK3P Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of GK3P.

Recommended Dilutions: Western Blot: 1:500-1:1000; ELISA: 1:40000

Type: Primary
Antigen: GK3P
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR