Anti-CNTF Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB18802
ABNOPAB18802EA 423 EUR
ABNOPAB18802
Anti-CNTF Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of CNTF.

Recommended Dilutions: ELISA: 1:40000-1:80000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: CNTF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR