Anti-CNTFR Rabbit Polyclonal Antibody
Lieferant: Abnova
PAB18804
ABNOPAB18804EA 423 EUR
ABNOPAB18804
Anti-CNTFR Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of CNTFR.
Recommended Dilutions: ELISA: 1:8000-1:30000; Western Blot: 1:200-1:500
Type: Primary
Antigen: CNTFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human