Anti-ESR2 Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB18806
ABNOPAB18806EA 423 EUR
ABNOPAB18806
Anti-ESR2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of ESR2.

Recommended Dilutions: ELISA: 1:40000-1:80000; Western Blot: 1:200-1:1000

Type: Primary
Antigen: ESR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR