Anti-ACTG1 Rabbit Polyclonal Antibody
Lieferant: Abnova
PAB18807
ABNOPAB18807EA 423 EUR
ABNOPAB18807
Anti-ACTG1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of ACTG1.
Recommended Dilutions: ELISA: 1:16000-1:80000; Western Blot: 1:300-1:500
Type: Primary
Antigen: ACTG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse