Anti-ADAMTS8 Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB18816
ABNOPAB18816EA 423 EUR
ABNOPAB18816
Anti-ADAMTS8 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of ADAMTS8.

Recommended Dilutions: ELISA: 1:80000; Western Blot: 1:500-1:1000

Type: Primary
Antigen: ADAMTS8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR