Anti-CASP7 Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB18850
ABNOPAB18850EA 423 EUR
ABNOPAB18850
Anti-CASP7 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of CASP7.

Recommended Dilutions: ELISA: 1:5000-1:20000; Immunohistochemistry: 1:50-1:200; Immunofluorescence: 1:50-1:100

Type: Primary
Antigen: CASP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR