Anti-ITIH4 Goat Polyclonal Antibody

Lieferant: Abnova
PAB19476
ABNOPAB19476EA 523 EUR
ABNOPAB19476
Anti-ITIH4 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of ITIH4.

Recommended Dilutions: ELISA: 1:1600; Western Blot: 0.03-0.1 ?g/ml

Type: Primary
Antigen: ITIH4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR