Anti-GRHL1 Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB20348
ABNOPAB20348EA 523 EUR
ABNOPAB20348
Anti-GRHL1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant GRHL1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500

Type: Primary
Antigen: GRHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR