Anti-PAWR Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB20610
ABNOPAB20610EA 523 EUR
ABNOPAB20610
Anti-PAWR Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant PAWR.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: PAWR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR