Anti-AQP1 Rabbit Polyclonal Antibody
Lieferant: Abnova
PAB27167
ABNOPAB27167EA 523 EUR
ABNOPAB27167
Anti-AQP1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of AQP1.
Recommended Dilutions: Western Blot: 0.1-0.5 ?g/ml ; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 0.5-1 ?g/ml
Type: Primary
Antigen: AQP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat