Anti-BAZ1A Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB28595
ABNOPAB28595EA 523 EUR
ABNOPAB28595
Anti-BAZ1A Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant BAZ1A.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: BAZ1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR