Anti-APP Rabbit Polyclonal Antibody
Lieferant: Abnova
PAB28928
ABNOPAB28928EA 779 EUR
ABNOPAB28928
Anti-APP Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody against amyloid oligomers.
  • Recommended Dilutions: Immunohistochemistry: 1:1000-10000; Immunoprecipitation: 1:1000; Western Blot
Type: Primary
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen