Anti-APP Rabbit Polyclonal Antibody (ATTO 390)
Lieferant: Abnova
PAB28929
ABNOPAB28929EA 912 EUR
ABNOPAB28929
Anti-APP Rabbit Polyclonal Antibody (ATTO 390)
Antikörper
Rabbit polyclonal antibody against amyloid oligomers.
Recommended Dilutions: Immunohistochemistry: 1:1000-10000; Immunoprecipitation: 1:1000; Western Blot
Type:
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: ATTO 390
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen