Anti-HSP90AB1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Abnova
PAB28955
ABNOPAB28955EA 648 EUR
ABNOPAB28955
Anti-HSP90AB1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of Hsp90b1.

Recommended Dilutions: Immunocytochemistry Immunohistochemistry Immunoprecipitation: 1:80; Western Blot: 1:1000

Type:
Antigen: HSP90AB1
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR