Anti-IgM Chicken Polyclonal Antibody

Lieferant: Abnova
PAB29187
ABNOPAB29187EA 402 EUR
ABNOPAB29187
Anti-IgM Chicken Polyclonal Antibody
Antikörper
Chicken polyclonal antibody raised against native Human IgM.

Type: Primary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR