Anti-c-Myc tag Chicken Polyclonal Antibody

Lieferant: Abnova
PAB29202
ABNOPAB29202EA 402 EUR
ABNOPAB29202
Anti-c-Myc tag Chicken Polyclonal Antibody
Antikörper
Chicken polyclonal antibody raised against c-Myc tag.

Recommended Dilutions: ELISA: Capture: 1:50-1:200, Detection: 1:5000-1:20000; Flow Cytometry: 1:100; Western Blot: 1:5000

Type: Primary
Antigen: c-Myc tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR