Anti-Rotavirus Goat Polyclonal Antibody

Lieferant: Abnova
PAB29808
ABNOPAB29808EA 402 EUR
ABNOPAB29808
Anti-Rotavirus Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against rotavirus.

Type: Primary
Antigen: Rotavirus
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Viruses
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR