Anti-Rubella virus Goat Polyclonal Antibody

Lieferant: Abnova
PAB29809
ABNOPAB29809EA 402 EUR
ABNOPAB29809
Anti-Rubella virus Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against rubella virus.

Type: Primary
Antigen: Rubella virus
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rubella virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR