Anti-Shigella Rabbit Polyclonal Antibody
Lieferant: Abnova
PAB29812
ABNOPAB29812EA 402 EUR
ABNOPAB29812
Anti-Shigella Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against Shigella.
Type: Primary
Antigen: Shigella
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria