Anti-Bacillus Rabbit Polyclonal Antibody
Lieferant: Abnova
PAB29827
ABNOPAB29827EA 402 EUR
ABNOPAB29827
Anti-Bacillus Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against Bacillus.
Type: Primary
Antigen: Bacillus
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria