Anti-FTCD Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB30090
ABNOPAB30090EA 523 EUR
ABNOPAB30090
Anti-FTCD Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of human FTCD.

Type: Primary
Antigen: FTCD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR