Anti-GMPPB Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB30104
ABNOPAB30104EA 523 EUR
ABNOPAB30104
Anti-GMPPB Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of human GMPPB.

Type: Primary
Antigen: GMPPB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR