Anti-GNAS Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB30105
ABNOPAB30105EA 523 EUR
ABNOPAB30105
Anti-GNAS Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of human GNAS.

Type: Primary
Antigen: GNAS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR