Anti-PKD2 Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB30512
ABNOPAB30512EA 523 EUR
ABNOPAB30512
Anti-PKD2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against partial recombinant human PKD2.

Type: Primary
Antigen: PKD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR