Anti-DUSP10 Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB30515
ABNOPAB30515EA 523 EUR
ABNOPAB30515
Anti-DUSP10 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against partial recombinant human DUSP10.

Type: Primary
Antigen: DUSP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR