Anti-CASP6 Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB30528
ABNOPAB30528EA 523 EUR
ABNOPAB30528
Anti-CASP6 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant human CASP6.

Type: Primary
Antigen: CASP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR