Anti-KIAA0323 Rabbit Polyclonal Antibody
Lieferant: Abnova
PAB30857
ABNOPAB30857EA 523 EUR
ABNOPAB30857
Anti-KIAA0323 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against partial recombinant human KIAA0323.
Type: Primary
Antigen: KIAA0323
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human