Anti-SNRPN Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB30861
ABNOPAB30861EA 523 EUR
ABNOPAB30861
Anti-SNRPN Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against partial recombinant human SNRPN.

Type: Primary
Antigen: SNRPN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR