Anti-HLA-DPA1 Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB30895
ABNOPAB30895EA 523 EUR
ABNOPAB30895
Anti-HLA-DPA1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against partial recombinant human HLA-DPA1.

Type: Primary
Antigen: HLA-DPA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR