Anti-NOS1AP Goat Polyclonal Antibody

Lieferant: Abnova
PAB6226
ABNOPAB6226EA 523 EUR
ABNOPAB6226
Anti-NOS1AP Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of NOS1AP.

Recommended Dilutions: ELISA: 1:8000; Immunohistochemistry: 5 ?g/ml

Type: Primary
Antigen: NOS1AP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR