Anti-EZR Goat Polyclonal Antibody

Lieferant: Abnova
PAB7060
ABNOPAB7060EA 523 EUR
ABNOPAB7060
Anti-EZR Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of EZR.

Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 3-5 ?g/ml

Type: Primary
Antigen: EZR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR