Anti-Amikacin Polyclonal Antibody

Lieferant: Abnova
PAB8219
ABNOPAB8219EA 1170 EUR
ABNOPAB8219
Anti-Amikacin Polyclonal Antibody
Antikörper
Sheep polyclonal antibody raised against Amikacin.

Recommended Dilutions: ELISA: 0.31 ?g/ml

Type: Primary
Antigen: Amikacin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR