Anti-DIG Polyclonal Antibody

Lieferant: Abnova
PAB8263
ABNOPAB8263EA 1170 EUR
ABNOPAB8263
Anti-DIG Polyclonal Antibody
Antikörper
Sheep polyclonal antibody raised against Digoxigenin.

Recommended Dilutions: ELISA: 0.625 ?g/ml

Type: Primary
Antigen: DIG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR