Anti-MPA Polyclonal Antibody

Lieferant: Abnova
PAB8297
ABNOPAB8297EA 1170 EUR
ABNOPAB8297
Anti-MPA Polyclonal Antibody
Antikörper
Sheep polyclonal antibody raised against Medroxyprogesterone acetate(3).

Recommended Dilutions: Enzyme Immunoassay (0.1 ?g/ml

Type: Primary
Antigen: MPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR