Anti-Nortriptyline Polyclonal Antibody

Lieferant: Abnova
PAB8330
ABNOPAB8330EA 1170 EUR
ABNOPAB8330
Anti-Nortriptyline Polyclonal Antibody
Antikörper
Sheep polyclonal antibody raised against Nortriptyline.

Recommended Dilutions: Enzyme Immunoassay (1.25 ?g/ml

Type: Primary
Antigen: Nortriptyline
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR