Anti-PCP(piperidyl-4) Polyclonal Antibody

Lieferant: Abnova
PAB8341
ABNOPAB8341EA 1170 EUR
ABNOPAB8341
Anti-PCP(piperidyl-4) Polyclonal Antibody
Antikörper
Sheep polyclonal antibody raised against PCP(piperidyl-4).

Recommended Dilutions: Enzyme Immunoassay (0.2 ?g/ml

Type: Primary
Antigen: PCP(piperidyl-4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR