Anti-Salbutamol Polyclonal Antibody

Lieferant: Abnova
PAB8355
ABNOPAB8355EA 912 EUR
ABNOPAB8355
Anti-Salbutamol Polyclonal Antibody
Antikörper
Sheep polyclonal antibody raised against Salbutamol.

Recommended Dilutions: Enzyme Immunoassay (5.75 ?g/ml

Type: Primary
Antigen: Salbutamol
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR