Anti-Sulphamerazine Polyclonal Antibody

Lieferant: Abnova
PAB8369
ABNOPAB8369EA 912 EUR
ABNOPAB8369
Anti-Sulphamerazine Polyclonal Antibody
Antikörper
Sheep polyclonal antibody raised against Sulphamerazine.

Recommended Dilutions: Enzyme Immunoassay (2.5 ?g/ml

Type: Primary
Antigen: Sulphamerazine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR