Anti-Tylosin Polyclonal Antibody

Lieferant: Abnova
PAB8393
ABNOPAB8393EA 1170 EUR
ABNOPAB8393
Anti-Tylosin Polyclonal Antibody
Antikörper
Sheep polyclonal antibody raised against Tylosin.

Recommended Dilutions: Enzyme Immunoassay (5 ?g/ml

Type: Primary
Antigen: Tylosin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR