Anti-Virginiamycin M Polyclonal Antibody

Lieferant: Abnova
PAB8394
ABNOPAB8394EA 1170 EUR
ABNOPAB8394
Anti-Virginiamycin M Polyclonal Antibody
Antikörper
Sheep polyclonal antibody raised against Virginiamycin M.

Recommended Dilutions: Enzyme Immunoassay (0.156 ?g/ml

Type: Primary
Antigen: Virginiamycin M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR