Anti-kay Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB8948
ABNOPAB8948EA 523 EUR
ABNOPAB8948
Anti-kay Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of kay.

Recommended Dilutions: Immunoprecipitation: 1:250; Immunohistochemistry: 1:200

Type: Primary
Antigen: kay
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Drosophila
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR