Anti-DBH Sheep Polyclonal Antibody

Lieferant: Abnova
PAB9692
ABNOPAB9692EA 523 EUR
ABNOPAB9692
Anti-DBH Sheep Polyclonal Antibody
Antikörper
Sheep polyclonal antibody raised against synthetic peptide of DBH.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: DBH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human, Mouse, Primates
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR