Anti-MLANA Monoclonal Antibody [clone: MLANA/1761R]

Lieferant: Abnova
RAB00275
ABNORAB00275EA 585 EUR
ABNORAB00275
Anti-MLANA Monoclonal Antibody [clone: MLANA/1761R]
Antikörper
Recombinant rabbit monoclonal antibody raised against human MLANA.

Type: Primary
Antigen: MLANA
Clonality: Monoclonal
Clone: MLANA/1761R
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR