Anti-EPCAM Monoclonal Antibody [clone: rMOC-31]

Lieferant: Abnova
RAB00303
ABNORAB00303EA 585 EUR
ABNORAB00303
Anti-EPCAM Monoclonal Antibody [clone: rMOC-31]
Antikörper
Recombinant mouse monoclonal antibody raised against human EPCAM.

Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: rMOC-31
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR