Anti-MS4A1 Monoclonal Antibody [clone: rIGEL/773]

Lieferant: Abnova
RAB00311
ABNORAB00311EA 585 EUR
ABNORAB00311
Anti-MS4A1 Monoclonal Antibody [clone: rIGEL/773]
Antikörper
Recombinant mouse monoclonal antibody raised against human MS4A1.

Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: rIGEL/773
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR