Anti-S1 protein hybridoma polyclonal antibody(ASS8)

Lieferant: ACROBIOSYSTEMS
S1N-S58-1MG S1N-S58-100UG
ACRBS1N-S58-1MGEA 2865 EUR
ACRBS1N-S58-1MG ACRBS1N-S58-100UG
Anti-S1 protein hybridoma polyclonal antibody(ASS8)
Antikörper
Type: Primary
Antigen: S1 protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hybridoma
Isotype: Mouse IgG1/kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR